Assay Details
Target Gene Details
Entrez Gene ID: | 231655 |
Gene Name: | 2'-5' oligoadenylate synthetase-like 1 |
Gene Aliases: |
7530414C13Rik, oasl9 |
Location: |
Chr.5:114923190-114937912 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oasl1 | NM_145209.3 | 2 | 151 | NP_660210.1 |
| AB067533.1 | 1 | 76 | BAB84133.1 | |
| AF426289.1 | 1 | 82 | AAN31518.1 | |
| AK078690.1 | 2 | 152 | BAC37360.1 | |
| AK089111.1 | 1 | 121 | ||
| AK149824.1 | 1 | 75 | BAE29106.1 | |
| AK150863.1 | 1 | 205 | BAE29916.1 | |
| AK165578.1 | 1 | 188 | BAE38269.1 | |
| AY057107.1 | 1 | 64 | AAL12828.1 | |
| AY089728.1 | 1 | 76 | AAM08092.1 | |
| BC032152.1 | 1 | 88 | AAH32152.1 |
More Information
Set Membership: |
|

Genomic Map