Assay Details
Target Gene Details
Entrez Gene ID: | 23962 |
Gene Name: | 2'-5' oligoadenylate synthetase-like 2 |
Gene Aliases: |
M1204, Mmu-OASL, Oasl |
Location: |
Chr.5:114896934-114912245 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oasl2 | NM_011854.2 | 4 | 1599 | NP_035984.2 |
| AF068835.1 | 4 | 1585 | AAD02818.1 | |
| AK010034.1 | 4 | 941 | BAB26655.1 | |
| AK150080.1 | 2 | 390 | BAE29289.1 | |
| AK150492.1 | 4 | 930 | BAE29608.1 | |
| AK157649.1 | 4 | 967 | ||
| AK160630.1 | 3 | 503 | ||
| BC034361.1 | 4 | 1026 | AAH34361.1 |
More Information
Set Membership: |
|

Genomic Map