Assay Details
Target Gene Details
Entrez Gene ID: | 72772 |
Gene Name: | RAD50 interactor 1 |
Gene Aliases: |
1500019C06Rik, 2810450M21Rik, Rint-1 |
Location: |
Chr.5:23787681-23820369 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rint1 | NM_177323.3 | NP_796297.2 | ||
| AK028707.1 | BAC26077.1 | |||
| AK036805.1 | BAC29582.1 | |||
| AK133789.1 | ||||
| AK135334.1 | BAE22492.1 | |||
| AK144144.1 | ||||
| BC065059.1 | AAH65059.2 | |||
| BC145267.1 | ||||
| BC150745.1 |

Genomic Map