Assay Details
Target Gene Details
Entrez Gene ID: | 69815 |
Gene Name: | keratinocyte associated protein 3 |
Gene Aliases: |
2010001C09Rik, AW321088, Kcp3 |
Location: |
Chr.5:31251706-31253197 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtcap3 | NM_027221.3 | NP_081497.2 | ||
| AK007999.1 | BAB25399.1 | |||
| BC027812.1 | AAH27812.1 | |||
| BC107024.1 | AAI07025.1 | |||
| BC107025.1 | AAI07026.1 | |||
| BQ033219.1 | ||||
| CK129285.1 |

Genomic Map