Assay Details
Target Gene Details
Entrez Gene ID: | 56433 |
Gene Name: | vacuolar protein sorting 29 (S. pombe) |
Gene Aliases: |
2010015D08Rik, AW049835, PEP11 |
Location: |
Chr.5:122354377-122364981 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps29 | AB578904.1 | |||
| AB578909.1 |

Genomic Map