Assay Details
Target Gene Details
Entrez Gene ID: | 56433 |
Gene Name: | vacuolar protein sorting 29 (S. pombe) |
Gene Aliases: |
2010015D08Rik, AW049835, PEP11 |
Location: |
Chr.5:122354377-122364981 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps29 | NM_019780.1 | 2 | 193 | NP_062754.1 |
| AB578899.1 | ||||
| AB578900.1 | ||||
| AB578901.1 | ||||
| AB578902.1 | ||||
| AB578903.1 | ||||
| AB578904.1 | ||||
| AB578905.1 | ||||
| AB578906.1 | ||||
| AB578907.1 | ||||
| AB578908.1 | ||||
| AB578909.1 | ||||
| AB578910.1 | 2 | 176 | ||
| AB578911.1 | 2 | 196 | ||
| AF193794.1 | 2 | 193 | AAF04595.1 | |
| AK004103.1 | 3 | 210 | BAB23170.1 | |
| AK008248.1 | 3 | 213 | ||
| AK036426.1 | 2 | 181 | ||
| AK044468.1 | 1 | 2515 | ||
| AK150330.1 | 2 | 203 | BAE29472.1 | |
| BC005663.1 | 2 | 182 | AAH05663.1 |
More Information
Set Membership: |
|

Genomic Map