Assay Details
Target Gene Details
Entrez Gene ID: | 57434 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 2 |
Gene Aliases: |
4921524O04Rik, 8030409M04Rik, RAD51, RecA |
Location: |
Chr.5:25689814-25705797 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc2 | NM_020570.2 | NP_065595.2 | ||
| AK020192.1 | BAB32023.1 | |||
| AK085093.1 | ||||
| BC033417.1 | AAH33417.1 | |||
| Y17040.1 | CAA76599.1 |
More Information
Set Membership: |
|

Genomic Map