Assay Details
Target Gene Details
Entrez Gene ID: | 75387 |
Gene Name: | sirtuin 4 |
Gene Aliases: |
4930596O17Rik |
Location: |
Chr.5:115478006-115490050 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sirt4 | NM_001167691.1 | NP_001161163.1 | ||
| NM_133760.1 | NP_598521.1 | |||
| AK016400.1 | ||||
| BC022653.1 | AAH22653.1 |

Genomic Map