Assay Details
Target Gene Details
Entrez Gene ID: | 74570 |
Gene Name: | zinc finger with KRAB and SCAN domains 1 |
Gene Aliases: |
5930429A01Rik, 9130423L19Rik, 9230118B16Rik, AI646829, AI788588, C87323, KOX18, PHZ-37, ZNF139 |
Location: |
Chr.5:138085076-138107822 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zkscan1 | NM_029869.1 | NP_084145.1 | ||
| NM_133906.4 | NP_598667.2 | |||
| AK020039.1 | BAB31975.1 | |||
| AK028932.1 | BAC26200.1 | |||
| AK031759.1 | BAC27539.1 | |||
| AK036807.1 | BAC29584.1 | |||
| AK054323.1 | BAC35730.1 | |||
| AK131148.1 | BAD21398.1 | |||
| AK135999.1 | BAE22767.1 | |||
| AK154831.1 | ||||
| BC052441.1 | AAH52441.1 |
More Information
Set Membership: |
|

Genomic Map