Assay Details
Target Gene Details
Entrez Gene ID: | 75750 |
Gene Name: | solute carrier family 10 (sodium/bile acid cotransporter family), member 6 |
Gene Aliases: |
8430417G17Rik, C78479, Soat |
Location: |
Chr.5:103605707-103629403 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc10a6 | NM_029415.2 | NP_083691.1 | ||
| AJ583504.1 | CAE47479.1 | |||
| AK018423.1 | BAB31203.1 | |||
| BC127608.1 | ||||
| BC139343.1 | ||||
| BC139346.1 |
More Information
Set Membership: |
|

Genomic Map