Assay Details
Target Gene Details
Entrez Gene ID: | 50878 |
Gene Name: | stromal antigen 3 |
Gene Aliases: |
SA-2 |
Location: |
Chr.5:138272589-138312397 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stag3 | NM_016964.2 | NP_058660.2 | ||
| AJ005678.1 | CAA06669.1 | |||
| AK145898.1 | BAE26734.1 | |||
| AK158240.1 | BAE34419.1 | |||
| BC139010.1 | ||||
| BC139011.1 |
More Information
Set Membership: |
|

Genomic Map