Assay Details
Target Gene Details
Entrez Gene ID: | 56334 |
Gene Name: | transmembrane emp24 domain trafficking protein 2 |
Gene Aliases: |
1110032D12Rik, 1810020N21Rik, Rnp24, Sid394, p24beta1 |
Location: |
Chr.5:124540791-124550503 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmed2 | NM_019770.2 | NP_062744.1 | ||
| AB025218.1 | BAA84689.1 | |||
| AK089965.1 | BAC41019.1 | |||
| AK179651.1 | ||||
| AK180184.1 | ||||
| BC083140.1 | AAH83140.1 | |||
| BC131977.1 | ||||
| BC138294.1 |
More Information
Set Membership: |
|

Genomic Map