Assay Details
Target Gene Details
Entrez Gene ID: | 75732 |
Gene Name: | IQ motif containing D |
Gene Aliases: |
4933433C09Rik, AI662646 |
Location: |
Chr.5:120586060-120607243 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcd |
Target Gene Details
Entrez Gene ID: | 252972 |
Gene Name: | two pore channel 1 |
Gene Aliases: |
5730403B01Rik, Tpc1, mKIAA1169 |
Location: |
Chr.5:120534157-120588699 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpcn1 | NM_145853.2 | NP_665852.1 | ||
| AF217002.1 | AAG44100.1 | |||
| AK037856.1 | ||||
| AK077411.1 | BAE43360.1 | |||
| AK148129.1 | BAE28363.1 | |||
| BC058951.1 | AAH58951.1 |
More Information
Set Membership: |
|

Genomic Map