Assay Details
Target Gene Details
Entrez Gene ID: | 100647 |
Gene Name: | uroplakin 3B |
Gene Aliases: |
AI447729, AI957187, P35, PMS2L14, UpIIIb |
Location: |
Chr.5:136038496-136044993 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Upk3b | NM_175309.4 | 3 | 609 | NP_780518.1 |
| AK042067.1 | 3 | 405 | BAC31149.1 | |
| AK052525.1 | 3 | 611 | BAC35025.1 | |
| AK157647.1 | 3 | 416 | ||
| AY233463.1 | 3 | 403 | AAO89508.1 | |
| BC061224.1 | 3 | 406 | AAH61224.1 | |
| BC120594.1 | 3 | 359 | ||
| BC137742.1 | 3 | 359 |

Genomic Map