Assay Details
Target Gene Details
Entrez Gene ID: | 22788 |
Gene Name: | zona pellucida glycoprotein 3 |
Gene Aliases: |
Zp-3 |
Location: |
Chr.5:135980105-135988624 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zp3 | NM_011776.1 | 8 | 1164 | NP_035906.1 |
| AK131934.1 | 8 | 1163 | BAE20883.1 | |
| BC099465.1 | 8 | 1149 | AAH99465.1 | |
| BC100745.1 | 8 | 1164 | AAI00746.1 | |
| BC103583.1 | 8 | 1164 | AAI03584.1 | |
| BC103584.1 | 8 | 1164 | AAI03585.1 | |
| BC103585.1 | 8 | 1164 | AAI03586.1 | |
| M20026.1 | 8 | 1164 | AAB18629.1 |

Genomic Map