Assay Details
Target Gene Details
Entrez Gene ID: | 68364 |
Gene Name: | RIKEN cDNA 0610030E20 gene |
Gene Aliases: |
1500031N04Rik, 2810411C16Rik, AI461894 |
Location: |
Chr.6:72347023-72353162 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 0610030E20Rik | NM_026696.1 | NP_080972.1 | ||
| AK002701.1 | BAB22295.1 | |||
| AK013079.2 | 1 | 356 | ||
| AK085662.1 | 1 | 377 | ||
| BB844069.1 | ||||
| BC064080.1 | ||||
| BC066053.1 | ||||
| BC117812.1 |

Genomic Map