Assay Details
Target Gene Details
Entrez Gene ID: | 20907 |
Gene Name: | syntaxin 1A (brain) |
Gene Aliases: |
HPC-1 |
Location: |
Chr.5:135023487-135051104 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stx1a | NM_016801.3 | NP_058081.2 | ||
| AK144399.1 | 6 | 636 | ||
| BC047133.1 | AAH47133.1 | |||
| BC100446.1 | AAI00447.1 | |||
| D45208.2 | BAA28865.2 |
More Information
Set Membership: |
|

Genomic Map