Assay Details
Target Gene Details
Entrez Gene ID: | 109552 |
Gene Name: | sorcin |
Gene Aliases: |
2210417O06Rik, 2900070H08Rik, Sor |
Location: |
Chr.5:8046078-8069314 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sri | NM_001080974.2 | NP_001074443.1 | ||
| NM_025618.3 | NP_079894.2 | |||
| AK008404.1 | BAB25652.1 | |||
| AK008783.1 | BAB25891.1 | |||
| AK008970.1 | BAB25997.1 | |||
| AK134747.1 | BAE22266.1 | |||
| AK146071.1 | BAE26876.1 | |||
| AK146911.1 | BAE27523.1 | |||
| AK168572.1 | BAE40442.1 | |||
| AK168792.1 | BAE40625.1 | |||
| AK168900.1 | BAE40716.1 | |||
| BC065790.1 | AAH65790.1 | |||
| BY708689.1 |
More Information
Set Membership: |
|

Genomic Map