Assay Details
Target Gene Details
Entrez Gene ID: | 58522 |
Gene Name: | tripartite motif-containing 54 |
Gene Aliases: |
4930486E09Rik, 4930566I02Rik, MURF, MURF-3, MuRF3, Rnf30 |
Location: |
Chr.5:31116612-31137626 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim54 | NM_021447.2 | NP_067422.1 | ||
| AF294790.1 | AAG03076.1 | |||
| AK016235.1 | 3 | 650 | ||
| BC107258.2 | AAI07259.1 | |||
| BC107259.2 | AAI07260.1 |
More Information
Set Membership: |
|

Genomic Map