Assay Details
Target Gene Details
Entrez Gene ID: | 22051 |
Gene Name: | thyroid hormone receptor interactor 6 |
Gene Aliases: |
- |
Location: |
Chr.5:137309899-137314487 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip6 | NM_011639.3 | NP_035769.1 | ||
| AF097511.1 | AAC72380.1 | |||
| AF116823.1 | AAD45984.1 | |||
| AK010435.1 | BAB26937.1 | |||
| AK146823.1 | ||||
| AK159387.1 | BAE35042.1 | |||
| AK166024.1 | BAE38527.1 | |||
| BC138685.1 | ||||
| BC138686.1 |
More Information
Set Membership: |
|

Genomic Map