Assay Details
Target Gene Details
Entrez Gene ID: | 20348 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C |
Gene Aliases: |
1110036B02Rik, SemE, Semae |
Location: |
Chr.5:17574816-17730268 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3c | NM_013657.5 | 1 | 77 | NP_038685.3 |
| AK034239.1 | 1 | 136 | BAC28643.1 | |
| AK078589.1 | 1 | 129 | ||
| BC066852.1 | 1 | 77 | AAH66852.1 | |
| X85994.1 | CAA59986.1 |
More Information
Set Membership: |
|

Genomic Map