Assay Details
Target Gene Details
Entrez Gene ID: | 21343 |
Gene Name: | TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
80kDa, AW549759, TAF(II)80, TAFII70, Taf2e, p80 |
Location: |
Chr.5:138178617-138187446 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf6 | NM_009315.3 | 6 | 586 | NP_033341.1 |
| AK154867.1 | 6 | 620 | ||
| AK185698.1 | ||||
| AK189374.1 | 3 | 206 | ||
| AK213626.1 | 3 | 206 | ||
| AK214579.1 | 3 | 206 | ||
| AK216752.1 | ||||
| BC058583.1 | 6 | 586 | AAH58583.1 | |
| D49439.1 | 5 | 519 | BAA08417.1 |
More Information
Set Membership: |
|

Genomic Map