Assay Details
Target Gene Details
Entrez Gene ID: | 56449 |
Gene Name: | Y box protein 3 |
Gene Aliases: |
Csda, Dpba, MSY3, MSY4, Yb2, dbpA, oxyR |
Location: |
Chr.6:131364858-131388450 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybx3 | NM_011733.2 | 7 | 881 | NP_035863.1 |
| NM_139117.2 | 8 | 1088 | NP_620817.2 | |
| AF246224.1 | 8 | 930 | AAF61741.1 | |
| AF248546.1 | 8 | 910 | AAG14418.1 | |
| AF248547.1 | 7 | 703 | AAG14419.1 | |
| AK003145.1 | 7 | 814 | ||
| AK076041.1 | 8 | 1027 | BAC36141.1 | |
| BC027785.1 | 7 | 892 | AAH27785.1 | |
| BC048242.1 | 8 | 1076 | AAH48242.1 | |
| BC062377.1 | 7 | 869 | AAH62377.1 | |
| D14485.1 | 7 | 869 | BAA03376.1 |

Genomic Map