Assay Details
Target Gene Details
Entrez Gene ID: | 13426 |
Gene Name: | dynein cytoplasmic 1 intermediate chain 1 |
Gene Aliases: |
DH IC-1, DHIC-1, DIC, Dncic1, IC74 |
Location: |
Chr.6:5725633-6028039 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dync1i1 | NM_001191023.1 | 9 | 1208 | NP_001177952.1 |
| NM_001191025.1 | 9 | 1157 | NP_001177954.1 | |
| NM_001191026.1 | 10 | 1184 | NP_001177955.1 | |
| NM_001191027.1 | 9 | 1124 | NP_001177956.1 | |
| NM_010063.4 | 10 | 1217 | NP_034193.2 | |
| AF063229.1 | 10 | 1051 | AAC33444.1 | |
| AF063230.1 | 9 | 991 | AAC33445.1 | |
| AK158589.1 | 10 | 1185 | BAE34570.1 | |
| BC051992.1 | 10 | 945 | AAH51992.1 | |
| GU992206.1 | 9 | 949 | ||
| GU992207.1 | 9 | 865 |
More Information
Set Membership: |
|

Genomic Map