Assay Details
Target Gene Details
Entrez Gene ID: | 11677 |
Gene Name: | aldo-keto reductase family 1, member B3 (aldose reductase) |
Gene Aliases: |
ALR2, AR, Ahr-1, Ahr1, Akr1b1, Aldor1, Aldr1 |
Location: |
Chr.6:34303930-34317489 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1b3 | NM_009658.3 | NP_033788.3 | ||
| AK028079.1 | ||||
| AK138000.1 | ||||
| AK139250.1 | BAE23933.1 | |||
| AK145392.1 | BAE26408.1 | |||
| AK149861.1 | BAE29130.1 | |||
| AK150197.1 | BAE29373.1 | |||
| AK153949.1 | BAE32272.1 | |||
| AK164682.1 | BAE37874.1 | |||
| AK170663.1 | BAE41944.1 | |||
| AK181972.1 | ||||
| AK186187.1 | ||||
| AK195753.1 | ||||
| AK205407.1 | ||||
| AK206515.1 | ||||
| AK207228.1 | ||||
| AK207346.1 | ||||
| AK207579.1 | ||||
| AK210404.1 | ||||
| AK210578.1 | ||||
| AK210866.1 | ||||
| AK211155.1 | ||||
| AK211229.1 | ||||
| AK211270.1 | ||||
| AK211569.1 | ||||
| AK212385.1 | ||||
| AK212386.1 | ||||
| AK212410.1 | ||||
| AK212510.1 | ||||
| AK212574.1 | ||||
| AK213617.1 | ||||
| AK213838.1 | ||||
| AK215229.1 | ||||
| AK217363.1 | ||||
| AK217744.1 | ||||
| AK217924.1 | ||||
| BC004725.1 | AAH04725.1 | |||
| BC021655.1 | AAH21655.1 | |||
| BC085310.1 | AAH85310.1 | |||
| BY749711.1 | ||||
| D32250.1 | BAA06980.1 | |||
| L39795.1 | AAA62176.1 | |||
| U29152.1 | AAA69958.1 |
More Information
Set Membership: |
|

Genomic Map