Assay Details
Target Gene Details
Entrez Gene ID: | 14080 |
Gene Name: | fatty acid binding protein 1, liver |
Gene Aliases: |
Fabpl, L-FABP |
Location: |
Chr.6:71199888-71205023 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fabp1 | NM_017399.4 | NP_059095.1 | ||
| AK131868.1 | BAE20841.1 | |||
| BC009812.1 | AAH09812.1 | |||
| Y14660.1 | CAA74989.1 |

Genomic Map