Assay Details
Target Gene Details
Entrez Gene ID: | 14187 |
Gene Name: | aldo-keto reductase family 1, member B8 |
Gene Aliases: |
FR-1, Fgfrp, Fgrp |
Location: |
Chr.6:34354164-34368454 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1b8 | NM_008012.1 | NP_032038.1 | ||
| BC005789.1 | AAH05789.1 | |||
| U04204.1 | AAA16953.1 |
More Information
Set Membership: |
|

Genomic Map