Assay Details
Target Gene Details
Entrez Gene ID: | 208665 |
Gene Name: | aldo-keto reductase family 1, member D1 |
Gene Aliases: |
- |
Location: |
Chr.6:37529978-37568815 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1d1 | NM_145364.2 | NP_663339.1 | ||
| BC018333.1 | AAH18333.1 |
More Information
Set Membership: |
|

Genomic Map