Assay Details
Target Gene Details
Entrez Gene ID: | 209773 |
Gene Name: | DENN/MADD domain containing 2A |
Gene Aliases: |
B930096L08Rik, C130031G22, mKIAA1277 |
Location: |
Chr.6:39462374-39557864 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd2a | NM_172477.4 | NP_766065.1 | ||
| AK048041.1 | ||||
| AK076686.1 | ||||
| AK081176.1 | BAC38157.1 | |||
| AK135781.1 | BAE22660.1 | |||
| AK160784.1 | BAE36008.1 | |||
| AK165202.1 | ||||
| AK167298.1 | BAE39403.1 | |||
| BC145599.1 | ||||
| BC150799.1 |

Genomic Map