Assay Details
Target Gene Details
Entrez Gene ID: | 209773 |
Gene Name: | DENN/MADD domain containing 2A |
Gene Aliases: |
B930096L08Rik, C130031G22, mKIAA1277 |
Location: |
Chr.6:39462374-39557864 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd2a | NM_172477.4 | 14 | 2779 | NP_766065.1 |
| AK048041.1 | 14 | 2764 | ||
| AK076686.1 | 6 | 922 | ||
| AK081176.1 | 14 | 2780 | BAC38157.1 | |
| AK135781.1 | 12 | 1324 | BAE22660.1 | |
| AK160784.1 | 14 | 2541 | BAE36008.1 | |
| AK165202.1 | 14 | 2634 | ||
| AK167298.1 | 14 | 2633 | BAE39403.1 | |
| BC145599.1 | 13 | 2458 | ||
| BC150799.1 | 13 | 2458 |

Genomic Map