Assay Details
Target Gene Details
Entrez Gene ID: | 76156 |
Gene Name: | family with sequence similarity 131, member B |
Gene Aliases: |
6330503C03Rik, 6530406I18Rik, AV277466 |
Location: |
Chr.6:42315305-42324640 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam131b | NM_001113327.2 | NP_001106798.1 | ||
| NM_001286584.1 | NP_001273513.1 | |||
| NM_029528.5 | NP_083804.3 | |||
| AK018206.1 | BAB31120.1 | |||
| AK018325.1 | ||||
| AK047763.1 | BAC33149.1 | |||
| AK158869.1 | BAE34703.1 | |||
| AK163654.1 | BAE37440.1 | |||
| BY260739.1 | ||||
| CJ101264.1 |

Genomic Map