Assay Details
Target Gene Details
Entrez Gene ID: | 12343 |
Gene Name: | capping protein (actin filament) muscle Z-line, alpha 2 |
Gene Aliases: |
1110053K06Rik, AW208808, Cappa2 |
Location: |
Chr.6:17637098-17666534 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capza2 | NM_007604.2 | NP_031630.1 | ||
| AK007550.1 | ||||
| AK008204.1 | ||||
| AK150587.1 | BAE29681.1 | |||
| AK150751.1 | BAE29821.1 | |||
| AK152674.1 | BAE31406.1 | |||
| AY544984.1 | AAS50156.1 | |||
| BC082589.1 | AAH82589.1 | |||
| U16741.1 | AAC00567.1 |
More Information
Set Membership: |
|

Genomic Map