Assay Details
Target Gene Details
Entrez Gene ID: | 12515 |
Gene Name: | CD69 antigen |
Gene Aliases: |
5830438K24Rik, AI452015, AIM, VEA |
Location: |
Chr.6:129267324-129275369 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd69 | NM_001033122.3 | NP_001028294.1 | ||
| AK017979.1 | ||||
| AK151076.1 | BAE30091.1 | |||
| AK153224.1 | BAE31817.1 | |||
| BC128272.1 | ||||
| BC128273.1 | ||||
| BC139135.1 | ||||
| BC139136.1 |

Genomic Map