Assay Details
Target Gene Details
Entrez Gene ID: | 12526 |
Gene Name: | CD8 antigen, beta chain 1 |
Gene Aliases: |
Cd8b, Ly-3, Ly-C, Lyt-3 |
Location: |
Chr.6:71322812-71337451 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd8b1 | NM_009858.2 | NP_033988.1 | ||
| AK087944.1 | BAC40052.1 | |||
| AK088372.1 | BAC40311.1 | |||
| AK088649.1 | BAC40477.1 | |||
| AK154055.1 | BAE32345.1 | |||
| AK162993.1 | BAE37149.1 | |||
| AK169581.1 | BAE41240.1 | |||
| M16799.1 | AAA39479.1 | |||
| M17534.1 | AAA39455.1 | |||
| M19504.1 | AAA39454.1 | |||
| U34882.1 | AAA92534.1 | |||
| X07698.1 | CAA30537.1 |
More Information
Set Membership: |
|

Genomic Map