Assay Details
Target Gene Details
Entrez Gene ID: | 71238 |
Gene Name: | succinate dehydrogenase complex assembly factor 3 |
Gene Aliases: |
0610005M07Rik, 4933430A16Rik, Acn9 |
Location: |
Chr.6:6956018-7039220 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdhaf3 | NM_001077713.1 | NP_001071181.1 | ||
| AK002229.1 | ||||
| AK004417.1 | BAE43178.1 | |||
| AK016986.1 | ||||
| AK028031.1 | BAC25710.1 | |||
| BC118927.1 | ||||
| BC118928.1 |

Genomic Map