Assay Details
Target Gene Details
Entrez Gene ID: | 68089 |
Gene Name: | actin related protein 2/3 complex, subunit 4 |
Gene Aliases: |
20kDa, 5330419I20Rik, AI327076, p20-Arc |
Location: |
Chr.6:113378113-113390447 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arpc4 | NM_001170485.1 | NP_001163956.1 | ||
| NM_001170486.1 | NP_001163957.1 | |||
| NM_026552.3 | NP_080828.1 | |||
| AJ278129.1 | CAC34583.1 | |||
| AK017993.1 | BAB31026.1 | |||
| AK018069.1 | BAB31059.1 | |||
| AK030840.1 | 1 | 3773 | ||
| AK039215.1 | BAC30279.1 | |||
| AK076248.1 | BAC36272.1 | |||
| AK077325.1 | BAC36751.1 | |||
| AK133626.1 | BAE21755.1 | |||
| AK136082.1 | BAE22810.1 | |||
| AK150690.1 | BAE29769.1 | |||
| AK153380.1 | BAE31946.1 | |||
| AK159410.1 | BAE35061.1 | |||
| AK166440.1 | BAE38777.1 | |||
| AK193553.1 | ||||
| AK194624.1 | ||||
| AK194784.1 | ||||
| AK197517.1 | ||||
| BC055309.1 | AAH55309.1 |
More Information
Set Membership: |
|

Genomic Map