Assay Details
Target Gene Details
Entrez Gene ID: | 14793 |
Gene Name: | cell division cycle associated 3 |
Gene Aliases: |
2410005A12Rik, C8, Grcc8, Tome-1 |
Location: |
Chr.6:124829595-124833701 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdca3 | NM_013538.5 | 2 | 175 | NP_038566.1 |
| AK008606.1 | 2 | 128 | BAB25773.1 | |
| AK010409.1 | 2 | 117 | BAB26916.1 | |
| AK011313.1 | 2 | 149 | BAB27539.1 | |
| AK028159.1 | 2 | 117 | BAC25785.1 | |
| AK145537.1 | 2 | 117 | BAE26492.1 | |
| AK169792.1 | 2 | 123 | BAE41370.1 | |
| AK211930.1 | 1 | 76 | ||
| BB860939.1 | 2 | 181 | ||
| BC002006.1 | 2 | 129 | AAH02006.1 | |
| BC027172.1 | 2 | 96 | AAH27172.1 |

Genomic Map