Assay Details
Target Gene Details
Entrez Gene ID: | 14793 |
Gene Name: | cell division cycle associated 3 |
Gene Aliases: |
2410005A12Rik, C8, Grcc8, Tome-1 |
Location: |
Chr.6:124829595-124833701 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdca3 | NM_013538.5 | NP_038566.1 | ||
| AK008606.1 | BAB25773.1 | |||
| AK010409.1 | BAB26916.1 | |||
| AK011313.1 | BAB27539.1 | |||
| AK028159.1 | BAC25785.1 | |||
| AK145537.1 | BAE26492.1 | |||
| AK169792.1 | BAE41370.1 | |||
| AK198356.1 | ||||
| BC002006.1 | AAH02006.1 | |||
| BC027172.1 | AAH27172.1 |

Genomic Map