Assay Details
Target Gene Details
Entrez Gene ID: | 269799 |
Gene Name: | C-type lectin domain family 4, member a1 |
Gene Aliases: |
BC049354, Dcir4 |
Location: |
Chr.6:122921848-122934619 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec4a1 | NM_199311.2 | 4 | 517 | NP_955015.1 |
| AY397675.1 | 4 | 452 | AAR31151.1 | |
| BC049354.1 | 4 | 507 | AAH49354.1 | |
| CK626014.1 | 4 | 517 |
More Information
Set Membership: |
|

Genomic Map