Assay Details
Target Gene Details
Entrez Gene ID: | 27999 |
Gene Name: | family with sequence similarity 3, member C |
Gene Aliases: |
D6Wsu176e, Ilei |
Location: |
Chr.6:22306520-22356081 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam3c | NM_138587.4 | NP_613053.3 | ||
| AK154498.1 | BAE32629.1 | |||
| AK170887.1 | BAE42094.1 | |||
| AY424275.1 | AAR84605.1 | |||
| BC009086.1 | AAH09086.1 | |||
| BC055853.1 | AAH55853.1 | |||
| BC069880.1 | AAH69880.1 |
More Information
Set Membership: |
|

Genomic Map