Assay Details
Target Gene Details
Entrez Gene ID: | 474145 |
Gene Name: | C-type lectin domain family 4, member a4 |
Gene Aliases: |
Dcir2 |
Location: |
Chr.6:122990367-123024421 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec4a4 | NM_001005860.2 | NP_001005860.1 | ||
| AY397673.1 | AAR31149.1 | |||
| BC104370.1 | AAI04371.1 |
More Information
Set Membership: |
|

Genomic Map