Assay Details
Target Gene Details
Entrez Gene ID: | 54722 |
Gene Name: | deafness, autosomal dominant 5 (human) |
Gene Aliases: |
2310037D07Rik, 4932441K13Rik, Dfna5h, EG14210, Fin15 |
Location: |
Chr.6:50207403-50262862 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dfna5 | NM_018769.3 | 8 | 1205 | NP_061239.1 |
| AF073309.1 | 8 | 1206 | AAC69325.1 | |
| AK171278.1 | 8 | 1204 | BAE42362.1 | |
| BC132303.1 | 7 | 1176 |

Genomic Map