Assay Details
Target Gene Details
Entrez Gene ID: | 74068 |
Gene Name: | ankyrin repeat, SAM and basic leucine zipper domain containing 1 |
Gene Aliases: |
4933400N19Rik, Gasz, ORF3 |
Location: |
Chr.6:18050964-18109061 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asz1 | NM_023729.3 | 5 | 518 | NP_076218.3 |
| AF459789.1 | 5 | 512 | AAL67487.1 | |
| AK016595.1 | 5 | 513 | ||
| BC058859.1 | 5 | 527 | AAH58859.1 | |
| BG088136.2 | 5 | 518 |

Genomic Map