Assay Details
Target Gene Details
Entrez Gene ID: | 12417 |
Gene Name: | chromobox 3 |
Gene Aliases: |
HP1g, M32 |
Location: |
Chr.6:51470616-51483704 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbx3 | NM_007624.3 | NP_031650.3 | ||
| AK002910.1 | BAB22450.1 | |||
| AK017958.1 | 1 | 107 | ||
| AK083957.1 | BAC39078.1 | |||
| AK146376.1 | BAE27125.1 | |||
| BC059831.1 | AAH59831.1 | |||
| BC108370.1 | AAI08371.1 | |||
| CJ093936.1 |

Genomic Map