Assay Details
Target Gene Details
Entrez Gene ID: | 214932 |
Gene Name: | cat eye syndrome chromosome region, candidate 5 |
Gene Aliases: |
A930002G03Rik |
Location: |
Chr.6:120509494-120531364 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cecr5 | NM_144815.2 | 6 | 662 | NP_659064.1 |
| AK140387.1 | 6 | 683 | ||
| BC014705.1 | 6 | 671 | AAH14705.1 |
More Information
Set Membership: |
|

Genomic Map