Assay Details
Target Gene Details
Entrez Gene ID: | 17474 |
Gene Name: | C-type lectin domain family 4, member d |
Gene Aliases: |
Clecsf8, Mpcl, mcl |
Location: |
Chr.6:123262107-123275268 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec4d | NM_001163161.1 | NP_001156633.1 | ||
| NM_010819.4 | NP_034949.3 | |||
| AF061272.1 | AAD05125.1 | |||
| AK089500.1 | BAC40905.1 | |||
| AK150926.1 | BAE29964.1 | |||
| AK155919.1 | ||||
| AK170692.1 | ||||
| AK171409.1 | BAE42436.1 | |||
| BC099471.1 | AAH99471.1 |
More Information
Set Membership: |
|

Genomic Map