Assay Details
Target Gene Details
Entrez Gene ID: | 56619 |
Gene Name: | C-type lectin domain family 4, member e |
Gene Aliases: |
C86253, Clecsf9, Mincle |
Location: |
Chr.6:123281789-123289871 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec4e | NM_019948.2 | NP_064332.1 | ||
| AB024717.1 | BAA83754.1 | |||
| AK036493.1 | ||||
| AK089286.1 | ||||
| AK150528.1 | BAE29637.1 | |||
| AK150899.1 | BAE29944.1 | |||
| AK151101.1 | BAE30111.1 | |||
| AK151912.1 | BAE30792.1 | |||
| AK152333.1 | ||||
| AK153008.1 | BAE31647.1 | |||
| AK153308.1 | BAE31889.1 | |||
| AK155452.1 | BAE33271.1 | |||
| AK156437.1 | BAE33711.1 | |||
| AK170698.1 | BAE41962.1 | |||
| AK171103.1 | BAE42251.1 | |||
| BC003218.1 | AAH03218.1 | |||
| CT010223.1 | CAJ18431.1 |
More Information
Set Membership: |
|

Genomic Map