Assay Details
Target Gene Details
Entrez Gene ID: | 319352 |
Gene Name: | PILR alpha associated neural protein |
Gene Aliases: |
AI255183, AU067746, C530028O21Rik, Panp |
Location: |
Chr.6:124996720-125003096 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pianp | NM_175696.4 | NP_783627.2 | ||
| AK082984.1 | BAC38721.1 | |||
| BB868634.1 |
More Information
Set Membership: |
|

Genomic Map