Assay Details
Target Gene Details
Entrez Gene ID: | 12638 |
Gene Name: | cystic fibrosis transmembrane conductance regulator |
Gene Aliases: |
AW495489, Abcc7 |
Location: |
Chr.6:18170262-18322774 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cftr | NM_021050.2 | 20 | 3374 | NP_066388.1 |
| M60493.1 | 20 | 3375 | AAA18903.1 | |
| M69298.1 | 20 | 3373 | AAA37417.1 |
More Information
Set Membership: |
|

Genomic Map