Assay Details
Target Gene Details
Entrez Gene ID: | 320609 |
Gene Name: | striatin interacting protein 2 |
Gene Aliases: |
D330017J20Rik, Fam40b |
Location: |
Chr.6:29916976-29959681 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strip2 | NM_001037740.1 | 9 | 983 | NP_001032829.1 |
| NM_177204.3 | 9 | 983 | NP_796178.2 | |
| NR_104576.1 | 9 | 983 | ||
| AK041004.1 | 9 | 984 | ||
| AK042073.1 | 9 | 984 | BAC31154.1 | |
| AK052271.1 | 9 | 984 | ||
| AK052273.1 | ||||
| AK122459.1 | 9 | 788 | BAC65741.1 | |
| AK163706.1 | 9 | 983 | BAE37467.1 | |
| BC147502.1 | 9 | 975 |

Genomic Map